missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, unpolished, Thermo Scientific™

Catalog Number 15436205
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 7647-14-5
Quantity 1 Ea.
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Chemical Name or Material Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, Unpolished
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'