missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm, unpolished

Spectroscopy, IR window

Brand:  Alfa Aesar™ 038395.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15426205

  • £21.80 / Each


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

Sodium chloride crystal optic rectangle
30mm x 15mm x 4mm, Unpolished
1 each
Videos
SDS