missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium chloride crystal optic disc, 32mm x 3mm, unpolished

Brand:  Alfa Aesar™ 038385.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15456195

  • £13.60 / Each


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Specifications

Sodium chloride crystal optic disc
32mm x 3mm, Unpolished
1 each
Videos
SDS