missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride crystal optic disc, 13mm x 2mm, unpolished, Thermo Scientific™

Spectroscopy, IR window

Brand:  Thermo Scientific Alfa Aesar 038383.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15436195

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Sodium chloride crystal optic disc
13mm x 2mm, Unpolished
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers