missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride crystal optic disc, 13mm x 2mm, polished both sides, Thermo Scientific Chemicals

Catalog Number 15416215
28.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Disc
Quantity 1 Ea.
Chemical Name or Material Sodium chloride crystal, Polished on both sides

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'