missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15528154
127.17 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
sodium chloride
[Na+].[Cl-]
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
sodium chloride
[Na+].[Cl-]

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
Formula Weight 58.44
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.16kg
Chemical Name or Material Sodium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'