missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99+%, extra pure, ACROS Organics™

Brand:  Acros Organics 223220010

Code : 29

Additional Details : CAS Number : 7775-09-9 Weight : 1.05000kg Transport : UN number : 1495 Chem class : 1 Pack group : II

Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10697202

  • £31.10 / 1kg


Estimated Shipment
01-02-2018
Add to basket

Specifications

Chemical Identifiers

Sodium chlorate
Extra Pure
99+%
NaClO3
7775-09-9
≥99%
ClNaO3
106.44

Specifications

Sodium chlorate
Extra Pure
≥99%
ClNaO3
106.44
0.005% max.
1000ppm max.
2.49
White
1kg
7775-09-9
Crystals
99+%
NaClO3
2.4900g/mL
Plastic bottle
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin, 7.7g/L ethanol (20°C)
0.05% max.
248.0° to 261.0°C
Videos
SDS