missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99%, ACS reagent, ACROS Organics™

Brand:  Acros Organics 446411000

Code : A0

Additional Details : CAS Number : 7775-09-9 Weight : 0.10000kg Transport : UN number : 1495 Chem class : 1 Pack group : II

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 12367173

  • £15.60 / 100g


Estimated Shipment
15-02-2018
Add to basket

Specifications

Chemical Identifiers

Sodium chlorate
ACS Reagent Grade
ClNaO3
106.44
7775-09-9
99%
NaClO3

Specifications

Sodium chlorate
ACS Reagent Grade
99%
NaClO3
2.4900g/mL
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin, 7.7g/L ethanol (20°C)
248.0° to 261.0°C
7775-09-9
99%
ClNaO3
106.44
Plastic bottle
2.49
100g
Videos
SDS