missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99%, ACS reagent, ACROS Organics™

Brand:  Acros Organics 446415000

Code : A0

Additional Details : CAS Number : 7775-09-9 Weight : 0.50000kg Transport : UN number : 1495 Chem class : 5.1 Pack group : II

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 12377173

  • £45.30 / 500g


Estimated Shipment
12-07-2018
Add to basket

Specifications

Chemical Identifiers

Sodium chlorate
ACS Reagent Grade
ClNaO3
106.44
7775-09-9
99%
NaClO3

Specifications

Sodium chlorate
ACS Reagent Grade
99%
NaClO3
2.4900g/mL
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin, 7.7g/L ethanol (20°C)
248.0° to 261.0°C
7775-09-9
99%
ClNaO3
106.44
Plastic bottle
2.49
500g
Videos
SDS