missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Anhydrous, ULTREX™ Ultrapure Reagent, J.T. Baker™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Quantity | 25 g |
Formula Weight | 105.99 |
Grade | ULTREX™ Ultrapure Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 2.53kg |
Chemical Name or Material | Sodium Carbonate, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'