missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390889
213.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
10340
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O
This item is not returnable. View return policy
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
10340
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 500 g
IUPAC Name disodium;carbonate
Formula Weight 105.99
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Chemical Name or Material Sodium Carbonate, Anhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'