missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Anhydrous, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
192.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Granules |
Quantity | 500 g |
IUPAC Name | disodium carbonate |
Formula Weight | 105.99 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 2.53kg |
Chemical Name or Material | Sodium Carbonate, Anhydrous |
Safety and Handling
Product Identifier
- Sodium Carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium carbonate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'