missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Anhydrous, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 3604-05

Code : NEW

Additional Details : CAS Number : 497-19-8 Weight : 2.50000kg

Quantity 2.5kg
Packaging Wide Mouth Amber Glass Bottle

 View more versions of this product

Product Code. 13518460

  • £312.00 / 2.50kg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

Sodium Carbonate
Anhydrous
Na2CO3
497-19-8
Baker Analyzed™ ACS Reagent
105.99

Specifications

Sodium Carbonate
497-19-8
Granular
105.99
1L = 2.53kg
Anhydrous
Baker Analyzed™ ACS Reagent
Na2CO3
Wide Mouth Amber Glass Bottle
2.5kg
Videos