missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate ACS AR Granular, Macron Fine Chemicals™
226.10 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Glass
Specifications
CAS | 497-19-8 |
Physical Form | Granules |
Quantity | 500 g |
Molecular Formula | CNa2O3 |
MDL Number | MFCD00003494 |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
IUPAC Name | disodium;carbonate |
Molecular Weight (g/mol) | 105.99 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'