missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical S/2640/63

Code : 21

Additional Details : CAS Number : 7647-15-6 Weight : 3.00000kg

Quantity 3kg

 View more versions of this product

Product Code. 10000370

Quantity Price
1 £162.44 / 3kg
4 £138.34 / 3kg
16 £131.41 / 3kg
In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Sodium Bromide, Extra Pure, SLR

Specifications

Chemical Identifiers

7647-15-6
102.89
JHJLBTNAGRQEKS-UHFFFAOYSA-M
253881
sodium;bromide
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
CHEBI:63004
[Na+].[Br-]

Specifications

7647-15-6
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium;bromide
253881
102.89g/mol
Plastic powder jar
White
5 to 8.8
BrNa
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
[Na+].[Br-]
102.89
CHEBI:63004
Powder Solid
1390°C
755°C
3kg
Videos
SDS Documents
Certificates