missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4782137
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
[Na+].[Br-]
This item is not returnable. View return policy
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
[Na+].[Br-]

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1390°C
Melting Point 755°C
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water and alcohol.
IUPAC Name sodium bromide
Formula Weight 102.89
Refractive Index 1.6412
Sensitivity Hygroscopic
Density 3.203 g/cm3 at 20°C
Chemical Name or Material Sodium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'