Learn More
Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 448510025
83.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 16940-66-2 Weight : 3.50000kg Transport : UN number : 3320 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
130.0°C to 135.0°C | |
VenPure™ solution, 12% aq. solution | |
2.5 L | |
NaBH4 | |
15,8727 | |
Solubility in water: completely soluble | |
[BH4-].[Na+] | |
37.83 | |
1.4 |
Sodium borohydride | |
16940-66-2,1310-73-2,67-56-1,7732-18-5 | |
BH4Na | |
MFCD00003518 | |
SBH, Sodium tetrahydroborate | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.82 | |
1.4000g/mL |
RUO – Research Use Only