Learn More
Sodium borohydride, 99%, powder, ACROS Organics™
Brand: Acros Organics 200051000
63.85 GBP valid until 2021-04-30
Use promo code "14803" to get your promotional price.
Code : 29
Additional Details : CAS Number : 16940-66-2 Weight : 0.15000kg Transport : UN number : 1426 Chem class : 4.3 Pack group : I
Packaging | Glass bottle |
---|---|
Quantity | 100g |
Description
Reduction reagentChemical Identifiers
16940-66-2 | |
H4BNa | |
MFCD00003518 | |
Sodium tetrahydroborate, SBH |
Specifications
Sodium Borohydride | |
Vapor Pressure: Negligible | |
H4BNa | |
MFCD00003518 | |
15, 8727 | |
37.83 | |
≥98.5% | |
Pass Test | |
>220°C | |
360°C |
16940-66-2 | |
99% | |
NaBH4 | |
UN 1426 | |
Sodium tetrahydroborate, SBH | |
Powder | |
1.3 | |
Glass bottle | |
White | |
100g |
Safety and Handling
- Sodium borohydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H301-Toxic if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
Precautionary Statement
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.