missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 99%, VenPure™ SF granules

Brand:  Acros Organics 448500100

Code : 29

Additional Details : CAS Number : 16940-66-2 Weight : 10.05000kg Transport : UN number : 1426 Chem class : 4.3 Pack group : I


 View more versions of this product

Product Code. 13276339

  • £1429.90 / 10kg


EU Stock 1
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Sodium borohydride
H4BNa
SBH, Sodium tetrahydroborate
16940-66-2
37.83

Specifications

Sodium borohydride
99%
H4BNa
37.83
Plastic drum
360.0°C
16940-66-2
99%
NaBH4
SBH, Sodium tetrahydroborate
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
10kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium borohydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H301-Toxic if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS