missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 98+%, powder, Acros Organics

Brand:  Acros Organics 189300025

Code : 29

Additional Details : CAS Number : 16940-66-2 Weight : 2.55000kg Transport : UN number : 1426 Chem class : 1 Pack group : I


Quantity 2.5kg
Packaging Plastic drum

 View more versions of this product

Product Code. 11934381

  • £590.80 / 2.50kg


EU Stock 3
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

Reduction reagent
Specifications

Chemical Identifiers

Sodium borohydride
Powder
98+%
NaBH4
MFCD00003518
16940-66-2
≥98%
H4BNa
37.83
SBH, Sodium tetrahydroborate

Specifications

Sodium borohydride
Vapor Pressure: Negligible
≥98%
H4BNa
37.83
01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
SBH, Sodium tetrahydroborate
Plastic drum
>220°C
360.0°C
16940-66-2
Powder
98+%
NaBH4
MFCD00003518
15, 8727
1.3
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
White
2.5kg
Videos
SDS