missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium borohydride, 20% aq. NaOH solution, ACROS Organics™
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Specifications
Quantity | 2.5 L |
Formula Weight | 37.83 |
Chemical Name or Material | Sodium borohydride, 20% aq. NaOH solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'