missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfate, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5390879
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
10034-88-5
H3NaO5S
138.07
MFCD00149210
JXHZRQHZVYDRGX-UHFFFAOYSA-M
sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
23673662
O.[Na+].OS([O-])(=O)=O
This item is not returnable. View return policy
10034-88-5
H3NaO5S
138.07
MFCD00149210
JXHZRQHZVYDRGX-UHFFFAOYSA-M
sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
23673662
O.[Na+].OS([O-])(=O)=O

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
IUPAC Name sodium;hydrogen sulfate;hydrate
Formula Weight 138.07
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Chemical Name or Material Sodium Bisulfate, Monohydrate
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'