missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfate, 92%, Tech., granular, Thermo Scientific Chemicals

Catalog Number p-3777063
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
7681-38-1
HNaO4S
120.05
MFCD00003531
WBHQBSYUUJJSRZ-UHFFFAOYSA-M
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
516919
sodium hydrogen sulfate
[Na+].OS([O-])(=O)=O
This item is not returnable. View return policy
7681-38-1
HNaO4S
120.05
MFCD00003531
WBHQBSYUUJJSRZ-UHFFFAOYSA-M
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
516919
sodium hydrogen sulfate
[Na+].OS([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 177.0°C to 180.0°C
Color White to Yellow
Physical Form Granules
Quantity 25 g
Assay Percent Range 92%
Linear Formula NaHSO4
Merck Index 15, 8722
Solubility Information Solubility in water: soluble. Other solubilities: decomposes by alcohol into na2so4 and h2so4
Formula Weight 120.06
Percent Purity 92%
Grade Technical
Packaging Glass bottle
Specific Gravity 2.1
Density 2.1000g/mL
Chemical Name or Material Sodium bisulfate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium bisulfate, tech., granular
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'