missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bismuthate, ACS reagent, Thermo Scientific Chemicals
44.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 g |
Linear Formula | NaBiO3 |
Merck Index | 15,8721 |
Solubility Information | Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids |
Formula Weight | 279.96 |
Grade | ACS Reagent |
Packaging | Glass bottle |
Chemical Name or Material | Sodium bismuthate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'