missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuth oxide, ACS, 80% min, Thermo Scientific Chemicals

Catalog Number p-7050003
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
12232-99-4
BiNaO3
279.967
MFCD00003474
PNYYBUOBTVHFDN-UHFFFAOYSA-N
sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
4063671
sodium;oxido(dioxo)bismuth
[O-][Bi](=O)=O.[Na+]
This item is not returnable. View return policy
12232-99-4
BiNaO3
279.967
MFCD00003474
PNYYBUOBTVHFDN-UHFFFAOYSA-N
sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
4063671
sodium;oxido(dioxo)bismuth
[O-][Bi](=O)=O.[Na+]

Manganese testing in iron, steel, ores

Sodium bismuth oxide is used for the determination of manganese in iron, steel and ores. It is also used in the separation of plutonium and acts as a strong oxidizer. It is used in the conversion of manganese compound to permanganate. Further, it is used in the preparation of hexavalent americium (Am) during its separation from the lanthanides and in the hydrothermal synthesis of bismuth oxychloride micrometer-sized single crystals. In addition to this, it is employed as a precursor in the preparation of bismuth nanoforms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Physical Form Powder
Quantity 25 g
Merck Index 14,8585
Solubility Information Insoluble in cold water; Decomposes in hot water forming Bi2O3,NaOH,and oxygenInsoluble in water.
Formula Weight 279.97
Percent Purity ≤80%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium bismuth oxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium bismuth oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'