missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
16.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Sodium bis(trifluoromethylsulfonyl)imide is used as an electrolyte in batteries and fuel cells. It also finds use as an ionic liquid and Lewis acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 257°C to 258°C |
Linear Formula | (CF3SO2)2NNa |
Quantity | 250 mg |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Formula Weight | 303.13 |
Physical Form | Solid |
Chemical Name or Material | Sodium bis (trifluoromethylsulfonyl)imide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'