missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate USP/FCC Powder, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 7396-06

Additional Details : Weight : 2.50000kg

Packaging Poly
Quantity 2.5kg

 View more versions of this product

Product Code. 15568414

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

144-55-8
84.006
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
C(=O)(O)[O-].[Na+]
CHNaO3
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium;hydrogen carbonate

Specifications

Sodium Bicarbonate
CHNaO3
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
84.01
FCC/USP
2.5kg
144-55-8
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
C(=O)(O)[O-].[Na+]
84.006
CHEBI:32139
Powder
Poly
Videos
Documents