missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate USP/FCC Powder, Macron Fine Chemicals™

Catalog Number p-3178124
85.03 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
This item is not returnable. View return policy
144-55-8
CHNaO3
84.01
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
516892
CHEBI:32139
sodium hydrogen carbonate
[Na+].OC([O-])=O
This item is not returnable. View return policy
144-55-8
CHNaO3
84.01
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
516892
CHEBI:32139
sodium hydrogen carbonate
[Na+].OC([O-])=O
Physical Form Powder
Quantity 500 g
Formula Weight 84.01
Grade FCC/USP
Packaging POLYSTORMOR
Chemical Name or Material Sodium Bicarbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'