missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | NaHCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4153970 |
Formula Weight | 84.01g/mol |
Percent Purity | 99 to 100.5% |
Grade | puriss. |
Chemical Name or Material | Sodium Bicarbonate, Meets analytical specification of Ph. Eur.,BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'