missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, Powder, U.S.P. - F.C.C., J.T.Baker™

Brand:  JT Baker 3509-05

Additional Details : CAS Number : 144-55-8 Weight : 3.00000kg

Packaging Wide Mouth Amber Glass Bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 15598134

  • £88.25 / 2.50kg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

144-55-8
84.006
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
C(=O)(O)[O-].[Na+]
CHNaO3
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium;hydrogen carbonate

Specifications

Sodium Bicarbonate
CHNaO3
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
84.01
FCC/USP
1L = 2.20kg
144-55-8
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
C(=O)(O)[O-].[Na+]
84.006
CHEBI:32139
Powder
Wide Mouth Amber Glass Bottle
2.5kg
Videos
Documents