missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™

Catalog Number p-46823
Click to view available options
Quantity:
12 kg
Packaging:
Poly Pail
This item is not returnable. View return policy
144-55-8
CHNaO3
84.01
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
516892
CHEBI:32139
sodium hydrogen carbonate
[Na+].OC([O-])=O
This item is not returnable. View return policy
144-55-8
CHNaO3
84.01
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
516892
CHEBI:32139
sodium hydrogen carbonate
[Na+].OC([O-])=O
Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 12 kg
Solubility Information Pass Test
Formula Weight 84.01
Percent Purity 99.0 to 101.0% (EP)
Grade BP/EP/FCC/JP/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/EP/BP/JP)
Loss on Drying 0.25% max. (FCC), 0.25% max. (USP), 0.25% max. (USP, FCC)
Packaging Poly Pail
Decomposition Information >50°C
Chemical Name or Material Sodium Bicarbonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'