missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, N.F. - F.C.C., J.T.Baker™

Brand:  JT Baker 3500-01

Additional Details : CAS Number : 532-32-1 Weight : 1.30000kg

Packaging Wide Mouth Amber Glass Bottle
Quantity 500g

 View more versions of this product

Product Code. 15538134

  • £80.29 / 500g

Estimated Shipment: 29-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

532-32-1
144.11
WXMKPNITSTVMEF-UHFFFAOYSA-M
517055
[Na+].[O-]C(=O)C1=CC=CC=C1
C7H5NaO2
MFCD00012463
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
sodium benzoate

Specifications

Sodium Benzoate
C7H5NaO2
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
[Na+].[O-]C(=O)C1=CC=CC=C1
144.11
144.11
Wide Mouth Amber Glass Bottle
500g
532-32-1
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate
517055
FCC/NF
1L = 1.44kg
Videos
SDS
Documents
Certificates