missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide OR, Macron Fine Chemicals™

Catalog Number p-3178111
Click to view available options
Quantity:
500 g
Packaging:
Poly
This item is not returnable. View return policy
26628-22-8
N3Na
65.011
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
This item is not returnable. View return policy
26628-22-8
N3Na
65.011
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
33557
CHEBI:278547
sodium;azide
[N-]=[N+]=[N-].[Na+]
Quantity 500 g
Formula Weight 65.01
Packaging Poly
Chemical Name or Material Sodium Azide
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'