missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Arsenite Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 2.5 L |
Packaging | HDPE plastic bottle |
Safety and Handling
Product Identifier
- Sodium arsenite solution 0.05M (0.1N)
- Danger
- Carcinogenicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 3
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'