Learn More
Alfa Aesar™ Sodium arsenite, 0.1N Standardized Solution
Brand: Alfa Aesar™ 041533.K7
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1686 Chem class : 1 Pack group : II
Quantity | 4L |
---|
Chemical Identifiers
Sodium arsenite | |
Standardized Solution | |
NaAsO2 | |
MFCD00003472 |
7784-46-5 | |
NaAsO2 | |
129.91 | |
UN1686 |
Specifications
Sodium arsenite | |
7784-46-5 | |
0.1N Standardized Solution | |
0.1N | |
129.91 | |
UN1686 | |
Colorless |
Standardized Solution | |
Liquid | |
NaAsO2 | |
NaAsO2 | |
MFCD00003472 | |
Odorless | |
4L |