missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals
18.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
Used in the laboratory as an essential ingredient of the microcosmic salt bead test for identification of metallic radicals. Also used as intermediates. Finds application in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 79°C (decomposition) |
Color | White |
Physical Form | Crystals |
Quantity | 100 g |
Solubility Information | Soluble in water. Insoluble in alcohol |
Formula Weight | 209.07 (137.02 Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Appearance | White crystals |
Density | 1.544 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'