missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium amide, 99%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 400.0°C |
Melting Point | 210.0°C |
Color | Gray-White to White |
Physical Form | Powder, Irregular Flakes or Granules |
Quantity | 100 g |
Assay Percent Range | 99% |
Linear Formula | NaNH2 |
Fieser | 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288 |
Merck Index | 15, 8714 |
Solubility Information | Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'