missing translation for 'onlineSavingsMsg'
Learn More

Sodium aluminium hydride, 93%, ACROS Organics™

Brand:  Acros Organics 380111000

Code : 29

Additional Details : CAS Number : 13770-96-2 Weight : 0.15000kg Transport : UN number : 2835 Chem class : 1 Pack group : II


Quantity 100g

 View more versions of this product

Product Code. 10320933

  • £237.20 / 100g


Estimated Shipment
15-02-2018
Add to basket


Specifications

Chemical Identifiers

13770-96-2
H4AlNa
MFCD00192443
93%
54

Specifications

Sodium aluminium hydride
93%
H4AlNa
MFCD00192443
Solubility in water: reacts violently.
100g
13770-96-2
93%
54
Glass bottle
178.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium aluminium hydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P223-Do not allow contact with water.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS