missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetylide, 18 wt.% slurry in xylene, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 10708888
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
1066-26-8, 1330-20-7
C2HNa
48.02
MFCD00008568
SFDZETWZUCDYMD-UHFFFAOYSA-N
sodium acetylide, sodium ethynide, ethynyl sodium, sodium acetylide suspension, sodium acetylide na c2h, sodium ethyne, monosodium acetylide, slbnqjagykqcki-uhfffaoysa-n, sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide
2733336
CHEBI:55387
sodium;ethyne
C#[C-].[Na+]
This item is not returnable. View return policy
1066-26-8, 1330-20-7
C2HNa
48.02
MFCD00008568
SFDZETWZUCDYMD-UHFFFAOYSA-N
sodium acetylide, sodium ethynide, ethynyl sodium, sodium acetylide suspension, sodium acetylide na c2h, sodium ethyne, monosodium acetylide, slbnqjagykqcki-uhfffaoysa-n, sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide
2733336
CHEBI:55387
sodium;ethyne
C#[C-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Physical Form Low-melting Solid or Crystalline Powder
Quantity 100 mL
Linear Formula HC≡CNa
Flash Point 25°C
Solubility Information Solubility in water: reacts
Formula Weight 48.02
Percent Purity 17.0 to 22.0 wt% (Total base)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.936
Density 0.9360g/mL
Chemical Name or Material Sodium acetylide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium acetylide, 18 wt% slurry in xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'