missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate trihydrate, Honeywell
Click to view available options
Quantity:
250 g
500 g
1 kg
2 kg
5 kg
Description
- ≥95% for general laboratory use.
Specifications
Quantity | 2 kg |
Linear Formula | CH3COONa · 3H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3732037 |
Formula Weight | 136.08g/mol |
Percent Purity | ≥99% |
Ignition Point | 1112°F |
Chemical Name or Material | Sodium acetate trihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'