missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™

Catalog Number 15166674
180.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
This item is not returnable. View return policy
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]
This item is not returnable. View return policy
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications
Physical Form Crystals
Quantity 1 kg
Formula Weight 136.08
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 1.45kg
Chemical Name or Material Sodium Acetate, Trihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'