missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390869
156.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]
This item is not returnable. View return policy
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
Formula Weight 136.08
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.45kg
Chemical Name or Material Sodium Acetate, Trihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'