missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Trihydrate (Colorless-to-white Crystals), Fisher BioReagents™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
Sodium Acetate Trihydrate (Colorless-to-white Crystals)
The trihydrate form of Sodium Acetate can be used for many of the same applications as the anhydrous form.
Specifications
Melting Point | 58°C |
Color | White |
pH | 8 to 9.5 |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | CH3COONa·3H2O |
Formula Weight | 136.08 |
Percent Purity | 99.0 to 101.0% |
Packaging | Poly Bottle |
Density | 1.45g/cm³ |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'