missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Trihydrate (Colorless-to-white Crystals), Fisher BioReagents™

Catalog Number 10091113
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]
This item is not returnable. View return policy
6131-90-4
C2H9NaO5
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
23665404
CHEBI:32138
sodium;acetate;trihydrate
CC(=O)[O-].O.O.O.[Na+]

Sodium Acetate Trihydrate (Colorless-to-white Crystals)

The trihydrate form of Sodium Acetate can be used for many of the same applications as the anhydrous form.
TRUSTED_SUSTAINABILITY
Melting Point 58°C
Color White
pH 8 to 9.5
Physical Form Solid
Quantity 500 g
Linear Formula CH3COONa·3H2O
Formula Weight 136.08
Percent Purity 99.0 to 101.0%
Packaging Poly Bottle
Density 1.45g/cm³
Chemical Name or Material Sodium Acetate Trihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'