Learn More
Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal, Fisher Chemical
Brand: Fisher Chemical S/2040/53
27.38 GBP valid until 2019-12-31
Use promo code "13870" to get your promotional price.
Code : 22
Additional Details : CAS Number : 6131-90-4 Weight : 0.50000kg
Packaging | Plastic powder jar |
---|---|
Quantity | 500g |
Chemical Identifiers
6131-90-4 | |
136.079 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
23665404 | |
sodium;acetate;trihydrate |
C2H9NaO5 | |
71557 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
Specifications
58°C | |
8 to 9.5 | |
White | |
C2H9NaO5 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
Solid |
500g | |
Plastic powder jar | |
6131-90-4 | |
71557 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
136.08g/mol |