missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Quantity | 500 g |
IUPAC Name | sodium;acetate |
Formula Weight | 82.03 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.53kg |
Chemical Name or Material | Sodium Acetate, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'