missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, 99+%, anhydrous, Molecular Biology Reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (1.19 g/L) at 20°C, and ethanol.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (1.19 g/L) at 20°C, and ethanol.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
Specifications
Melting Point | 324°C |
Color | White |
pH | 7.5 to 9 |
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,8571 |
IUPAC Name | sodium acetate |
Formula Weight | 82.03 |
Percent Purity | ≥99% |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'