missing translation for 'onlineSavingsMsg'
Learn More

Sodium for AAS 1g/L H2O matrix, Fisher chemical™

Catalog Number p-7083703
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
CHEBI:26708
sodium
[Na]
This item is not returnable. View return policy
7440-23-5
Na
22.99
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
CHEBI:26708
sodium
[Na]

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 1g/L ±0.002g/L
Packaging Bottle
Quantity 100 mL
Type Flame Single-element Standard Solution
Solvent or Matrix H2O
Physical Form Liquid
Chemical Name or Material Sodium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'