missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-nitrobenzenesulfonate, 98%, water <2%, Thermo Scientific Chemicals

Catalog Number p-7080633
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
127-68-4
C6H4NNaO5S
225.15
MFCD00007490
LJRGBERXYNQPJI-UHFFFAOYSA-M
sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
31389
sodium;3-nitrobenzenesulfonate
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]
This item is not returnable. View return policy
127-68-4
C6H4NNaO5S
225.15
MFCD00007490
LJRGBERXYNQPJI-UHFFFAOYSA-M
sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
31389
sodium;3-nitrobenzenesulfonate
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]

Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

Solubility
Soluble in water 200 g/L (20C).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 350°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water 200g/L (20C).
Formula Weight 225.16 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium 3-nitrobenzenesulfonate, Water <2%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'