Learn More
Alfa Aesar™ Sodium 3-(1-naphthylamino)propanesulfonate, 97%
Brand: Alfa Aesar™ H61707.06
Code : NEW
Additional Details : CAS Number : 104484-71-1 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Sodium 3-(1-naphthylamino)propanesulfonate | |
287.31 | |
3-(1-Naphthylamino)propanesulfonic acid sodium salt |
104484-71-1 | |
MFCD00059520 |
Specifications
Sodium 3-(1-naphthylamino)propanesulfonate | |
Solid | |
287.31 | |
3-(1-Naphthylamino)propanesulfonic acid sodium salt |
104484-71-1 | |
97% | |
MFCD00059520 | |
5g |